• USA Home
  • B7806 - Monoclonal Anti-Bcl-10 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
B7806 Sigma

Monoclonal Anti-Bcl-10 antibody produced in mouse

~2 mg/mL, clone 151, purified immunoglobulin, buffered aqueous solution

Properties

Related Categories AA - CA, Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology,
species reactivity   human
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections)1: suitable
  indirect ELISA1: suitable
  microarray: suitable
  western blot1, 2: 1-2 μg/mL using whole extract of cultured human lymphoma Raji cells.
clone   151, monoclonal
concentration   ~2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt ~32 kDa
  antigen mol wt ~37 kDa (weaker band)
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BCL10(8915)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant human Bcl-10.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Specificity

The epitope recognized by the antibody resides within amino acids 122-168 of human Bcl-10 molecule.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?