EMAIL THIS PAGE TO A FRIEND
B7886 Sigma

S-Butylglutathione

  • CAS Number 6803-16-3

  • Empirical Formula (Hill Notation) C14H25N3O6S

  • Molecular Weight 363.43

  •  MDL number MFCD00056733

  •  PubChem Substance ID 24892027

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Glutathione and Derivatives, Peptides
InChI Key   YGCGHOZSELIPKD-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?