• USA Home
  • B8125 - im-Benzyl-L-histidine

EMAIL THIS PAGE TO A FRIEND
B8125 Sigma

im-Benzyl-L-histidine

  • CAS Number 16832-24-9

  • Empirical Formula (Hill Notation) C13H15N3O2

  • Molecular Weight 245.28

  •  MDL number MFCD00038630

Purchase

Properties

InChI Key   NMXSWQMJOZUQKY-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?