• USA Home
  • B8256 - Bile acids, oxidized from ox or sheep

EMAIL THIS PAGE TO A FRIEND
B8256 Sigma

Bile acids, oxidized from ox or sheep

Purchase

Properties

solubility   methanol: soluble clear to hazy

Description

Other Notes

Predominantly dehydrocholic acid

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?