• USA Home
  • B8266 - Boc-O-benzyl-Ser-Gly-Arg-p-nitroanilide

EMAIL THIS PAGE TO A FRIEND
B8266 Sigma

Boc-O-benzyl-Ser-Gly-Arg-p-nitroanilide

  • CAS Number 77220-80-5

  • Empirical Formula (Hill Notation) C29H40N8O8

  • Molecular Weight 628.68

  •  MDL number MFCD00063346

  •  PubChem Substance ID 24892065

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   VVLVMZUTZYDQNL-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?