EMAIL THIS PAGE TO A FRIEND
B8406 Sigma

Bretylium tosylate

>99% (TLC), powder

Synonym: (o-Bromobenzyl)ethyldimethylammonium p-toluenesulfonate

Purchase

Properties

Related Categories Adrenergics, Cell Biology, Cell Signaling and Neuroscience, Neuroscience, Neurotransmission,
InChI Key   KVWNWTZZBKCOPM-UHFFFAOYSA-M
assay   >99% (TLC)
form   powder
color   white
solubility   H2O: 50 mg/mL

Description

Biochem/physiol Actions

Bretylium tosylate has a biphasic response, initially inducing norepinephrine release followed by sympathetic ganglionic blockade; class III antiarrhythmic agent; competitive inhibitor of acetylcholinesterase.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
BO9450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?