• USA Home
  • B8410 - Nalgene® bottles, style 2202
EMAIL THIS PAGE TO A FRIEND
B8410 Sigma
Nalgene® bottles, style 2202
size 4,000 mL
Synonym: nalgene bottle
Purchase
Properties
Related Categories Bottles and Caps, Labware, Low-Density Polyethylene Bottles, Manufacturer Browser, Nalgene,
material   low-density polyethylene carbonate bottle
  polypropylene screw closure
description   Not autoclavable
mfr. no.   Nalgene
size   4,000 mL
Description
General description
Narrow-mouth bottles. The 4 and 8 L sizes have a built-in shoulder loop.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?