• USA Home
  • B8807 - Biotin-β-D-glucopyranoside polyacrylamide

EMAIL THIS PAGE TO A FRIEND
B8807 Sigma

Biotin-β-D-glucopyranoside polyacrylamide

Synonym: β-D-Glc-PAA-biotin

Purchase

Description

Application

Biotin-β-D-glucopyranoside polyacrylamide may be used in the development of load-bearing hydrogels for tissue engineering applications.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?