• USA Home
  • B8811 - BLK, active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
B8811 Sigma

BLK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: MGC10442

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   95-129 nmol/min·mg
mol wt   mol wt ~84 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... BLK(640)

Description

Biochem/physiol Actions

BLK, also known as B lymphoid kinase, is a 55 kDa tyrosine kinase with SH3, SH2 and catalytic domains that contain consensus sequences of the src protein tyrosine kinase family. BLK is expressed specifically in the B cell lineage and plays a role in signal transduction pathway that is restricted to B lymphoid cells. Stimulation of resting B-lymphocytes with antibodies to surface immunoglobulin (sIgD or sIgM) induces activation of BLK (2)

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?