EMAIL THIS PAGE TO A FRIEND
B8888 Sigma

D-tert-Leucine

Synonym: (R)-2-Amino-3,3-dimethylbutyric acid, D-α-tert-Butylglycine

  • CAS Number 26782-71-8

  • Linear Formula (CH3)3CCH(NH2)CO2H

  • Molecular Weight 131.17

  •  Beilstein Registry Number 1721825

  •  MDL number MFCD00004265

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   NPDBDJFLKKQMCM-BYPYZUCNSA-N
mp   ≥300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

269115 D-tert-Leucine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?