• USA Home
  • B8932 - α-D-Mannopyranoside polyacrylamide biotin

EMAIL THIS PAGE TO A FRIEND
B8932 Sigma

α-D-Mannopyranoside polyacrylamide biotin

Synonym: α-D-Man-PAA-biotin

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Polyacrylamide (PAA)-based Glycoconjugates More...
mol wt   mol wt ~30 kDa

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?