EMAIL THIS PAGE TO A FRIEND
B9001 Sigma

Boc-Arg(Tos)-OH

Synonym: Nα-Boc-Nω-tosyl-L-arginine

  • CAS Number 13836-37-8

  • Empirical Formula (Hill Notation) C18H28N4O6S

  • Molecular Weight 428.50

  •  Beilstein Registry Number 2683538

  •  EC Number 237-549-2

  •  MDL number MFCD00065557

Purchase

Properties

InChI Key   WBIIPXYJAMICNU-AWEZNQCLSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

15506 Boc-Arg(Tos)-OH

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?