• USA Home
  • B9054 - Monoclonal Anti-Bax antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
B9054 Sigma

Monoclonal Anti-Bax antibody produced in mouse

2 mg/mL, clone 5B7, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Bcl-2 Family Proteins,
species reactivity   mouse
does not react with   human, rat
application(s)   immunoprecipitation: suitable
  microarray: suitable
  western blot: 1-5 μg/mL using mouse fibroblast 3T3 cell extract
clone   5B7, monoclonal
concentration   2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 21 kDa (and possibly a dimer of 42 kDa)
shipped in   dry ice
storage temp.   −20°C
Gene Information   mouse ... Bax(12028)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 3-16 of the mouse Bax sequence, conjugated to KLH

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Specificity

The epitope recognized by the antibody resides within amino acids 3-16 of the mouse Bax protein.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?