• USA Home
  • B9179 - Apoptosis PCR Primers

EMAIL THIS PAGE TO A FRIEND
B9179 Sigma

Apoptosis PCR Primers

Bcl-2 primers

Purchase

Properties

Related Categories Apoptosis Detection, Apoptosis and Cell Cycle, Cell Biology, Cell Signaling and Neuroscience, PCR Primer Sets More...
usage   sufficient for 50 PCR reactions
shipped in   wet ice
storage temp.   −20°C

Description

Application

Synthetic oligonucleotide primers for RT-PCR analysis of mRNA expression. Amplifies reverse-transcript mRNAs from human, mouse and rat sources.

Packaging

Set includes an upstream and a downstream oligonucleotide.

Price and Availability


Flow Cytometry Instruments
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?