EMAIL THIS PAGE TO A FRIEND
B9300 Sigma

Benzoic acid

meets USP testing specifications

  • CAS Number 65-85-0

  • Linear Formula C6H5COOH

  • Molecular Weight 122.12

  •  Beilstein Registry Number 636131

  •  EC Number 200-618-2

  •  MDL number MFCD00002398

  •  PubChem Substance ID 329773317

  •  eCl@ss 39023903

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Analytical/Chromatography, Benzoic Acid and Benzoic Acid Solutions, Building Blocks, C7, Carbonyl Compounds,
vapor density   4.21 (vs air)
vapor pressure   10 mmHg ( 132 °C)
InChI Key   WPYMKLBDIGXBTP-UHFFFAOYSA-N
autoignition temp.   1061 °F
bp   249 °C(lit.)
mp   121-125 °C(lit.)
suitability   meets USP testing specifications

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Lungs
Personal Protective Equipment 
WGK Germany 
1
RTECS 
DG0875000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?