• USA Home
  • B9303 - Monoclonal Anti-BAP1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
B9303 Sigma

Monoclonal Anti-BAP1 antibody produced in mouse

clone 3C11, ascites fluid

Synonym: Anti-BRCA1 Associated Protein 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Cycle, Antibodies to Nuclear Proteins,
species reactivity   human
application(s)   immunocytochemistry: suitable
  immunoprecipitation: suitable
  microarray: suitable
  western blot: 1:2,000 using a whole cell extract from a human melanoma cell line
clone   3C11, monoclonal
antibody form   ascites fluid
isotype   IgG1
mol wt   antigen mol wt 81 kDa
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BAP1(8314)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant full-length BAP1.

Specificity

The antibody recognizes an epitope within the region of amino acids 337-440 of the human BAP1 molecule. The antibody reacts with BAP1 (and additional lower and higher MW bands).

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?