• USA Home
  • B9502 - 8-Bromo-2′,3′,5′-tri-O-acetylguanosine

EMAIL THIS PAGE TO A FRIEND
B9502 Sigma

8-Bromo-2′,3′,5′-tri-O-acetylguanosine

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   JLZCAXCVNVVXJL-SYFKMIMOSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?