EMAIL THIS PAGE TO A FRIEND
B9507 Sigma

Myristyl behenate

Synonym: Behenic acid myristyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acyls, Fatty Esters, Lipids, Wax monoesters More...
InChI Key   AVKVDDQTHIQFSC-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?