EMAIL THIS PAGE TO A FRIEND
B9637 Sigma

Boc-Sar-OH

Synonym: Boc-N-methylglycine, Boc-sarcosine

  • CAS Number 13734-36-6

  • Linear Formula (CH3)3COCON(CH3)CH2COOH

  • Molecular Weight 189.21

  •  Beilstein Registry Number 2046827

  •  EC Number 237-306-0

  •  MDL number MFCD00037795

Purchase

Properties

InChI Key   YRXIMPFOTQVOHG-UHFFFAOYSA-N
mp   88-90 °C
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15495 Boc-Sar-OH, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?