• USA Home
  • BAF133520001 - Bel-Art® X-Ray Film Box Holder Bag
EMAIL THIS PAGE TO A FRIEND
BAF133520001 Sigma
Bel-Art® X-Ray Film Box Holder Bag
For 8 x 10 in. Cassettes
Purchase
Properties
Related Categories Autoradiography, Autoradiography Film Accessories, Electrophoresis Supplies, Labware
size   30 × 36 cm
Description
General description
These bags are ideal for shielding light sensitive X-ray films from ambient light especially those in storage boxes that have been spoiled.
Safeguards X-ray film from inadvertent light exposure in boxes that may be damaged
• Bag′s outer layer is black nylon with a tight weave
• Inner layer is made of light-blocking vinyl
• Large covering flap over bag opening is fastened by a hook-and-loop strip
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?