• USA Home
  • BE0200 - β Amyloid 1-42 ELISA human

EMAIL THIS PAGE TO A FRIEND
BE0200 Sigma

β Amyloid 1-42 ELISA human

sufficient for 96 multiwell tests

Synonym: β Amyloid ELISA, Amyloid β ELISA

Purchase

Properties

usage   sufficient for 96 multiwell tests
shipped in   wet ice
storage temp.   2-8°C

Description

Features and Benefits

The assay is a sensitive (10 pg/mL), and specific alternative to immunoblotting or RIA.

Application

Human β-amyloid (hAβ) ELISAs are designed for in vitro quantitative detection of native and recombinant β-amyloid in sera, cell culture supernatants and other biological samples. hAβ′s are main component of the protein deposits in neurofibrillary tangles, neuritic plaques and neuropil threads in the cerebral cortex of Alzheimer′s Disease (AD) and Down′s Syndrome. The antibodies used in these kits selectively differentiate between Aβ40 and Aβ42/Aβ43.

The ELISA can be used for in vitro quantitative determination of human native and recombinant β-amyloid 1-42 (hAβ42) protein in serum, cell culture supernatants and other biological samples. The Aβ42 antibody used in this kit selectively detects hAβ42 and not hAβ40.

General description

A solid-phase sandwich enzyme-linked immunosorbent assay (ELISA). A monoclonal capture antibody specific for the NH2 terminus of hAβ has been coated onto the multiwell strips. hAβ antigen in standards and samples binds to the capture antibody. A detection antibody specific for the hAβ 1−40 or 1−42 sequence binds the immobilized hAβ. An anti-rabbit IgG-HRP completes the sandwich. The reaction is visualized by TMB substrate, followed by the stop solution. The intensity of the yellow color is directly proportional to the concentration of hAβ in the original sample.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3316 9/PG 3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?