EMAIL THIS PAGE TO A FRIEND
BRAINPEP Sigma

Brain Peptide Kit

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
shipped in   wet ice
storage temp.   −20°C

Description

Application

The kit contains a group of tyrosine-containing peptides, four of which are related to met-enkephalin, that can be used for UV absorbance and fluorescence studies.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters

Kit component only

Description

Product #

Add to Cart

Tyr-D-Ala-Gly-Phe-Met-ol, solid SDS    
Tyr-D-Met-Gly-Phe-Pro-NH2, solid SDS    
Tyr-D-Ala-Gly-Phe-Met, solid SDS    
Tyr-D-Ala-Gly-Phe-Met(O)-ol, solid SDS    
Tyr-Phe-Arg, solid SDS    
Tyr-Ala-Arg acetate salt, solid SDS    
Tyr-Gly-Gly-Phe-Met, solid SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?