• USA Home
  • C0174 - Multiwell plate washer/dispenser manifolds

EMAIL THIS PAGE TO A FRIEND
C0174 Sigma

Multiwell plate washer/dispenser manifolds

Repeat kit

Purchase

Properties

Related Categories Drummond, Labware, Manufacturer Browser, Multiwell Plates, Washers, Orienters, etc. More...
sterility   non-sterile
color   none
matrix active group   Tissue culture treated surface treatment

Description

General description

Adapts manifolds for continuous delivery operation.
Includes 3-way valve with luer fittings, microvalve, stainless steel sinker, 30-inch length of 3/32 in. I.D. Tygon tubing, and tubing connectors.

For use with 96 well multiwell plates
• Dispense or aspirate media, or wash, a full row or column at a time
• Luer fitting
• Stainless steel and PTFE construction for easy cleaning and autoclavability
• Optional repeat kit provides continuous operation capability

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?