• USA Home
  • C0236 - Nunc® Lab-Tek® multi-purpose lab containers

EMAIL THIS PAGE TO A FRIEND
C0236 Sigma

Nunc® Lab-Tek® multi-purpose lab containers

size 8 oz, polypropylene

Purchase

Properties

Related Categories Containers, Labware, Manufacturer Browser, Nunc, Nunc Containers,
material   polypropylene
packaging   case of 120
mfr. no.   Nunc
diam. × H   114.3 mm × 44.5 mm
size   8 oz

Description

General description

• White containers with snug-fitting lids are designed for collection, transport and storage of lab specimens.
• Polypropylene containers are resistant to most chemicals and from freezing and boiling.
• Extra-large containers are polyethylene for added reliability and durability.

Legal Information

Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?