• USA Home
  • C0245 - CAMK4, active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
C0245 Sigma

CAMK4, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: CaMK-GR, MGC36771

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   78-106 nmol/min·mg
mol wt   mol wt ~79 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CAMK4(814)

Description

Biochem/physiol Actions

CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family. CAMK4 is localized in neurons in the hippocampus, amygdala, anterior cingulate cortex, somatosensory cortex, and insular cortex. CAMK4 is involved in neural activity-dependent signaling in the neuronal nucleus and thought to play an important role in the consolidation/retention of hippocampus-dependent long-term memory.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?