• USA Home
  • C0264 - Creatine hemisulfate salt

EMAIL THIS PAGE TO A FRIEND
C0264 Sigma

Creatine hemisulfate salt

Synonym: (α-Methylguanido)acetic acid, N-Amidinosarcosine

Purchase

Properties

Related Categories Cell Biology, Nutrition Research, Other Biochemical, Phytochemicals by Chemical Classification
InChI Key   NFWOMGOXOHCLNA-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?