EMAIL THIS PAGE TO A FRIEND
C0265 Sigma

Chromosorb® G

HP, 100-120 mesh particle size, bottle of 100 g

Purchase

Properties

InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
packaging   bottle of 100 g
application(s)   gas chromatography (GC): suitable
particle size   100-120 mesh
surface area   0.5 m2/g
density   0.47 g/cm3 (loose weight)(lit.)

Description

Preparation Note

Acid washed and treated with dimethyldichlorosilane

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?