• USA Home
  • C0487 - Clarion Mounting Medium

EMAIL THIS PAGE TO A FRIEND
C0487 Sigma

Clarion Mounting Medium

Purchase

Description

Application

Permanent, environmentally safe mounting medium for general use in Histology and Cytology. For use with insoluble organic chromogen substrates. Clarion is a trademerk of Biomeda Corp.

Legal Information

Clarion is a trademark of Biomeda Corp.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
136 °F
Flash Point(C) 
58 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

C9368 CC/Mount, tissue mounting medium

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?