EMAIL THIS PAGE TO A FRIEND
C0531 Sigma

Chlordiazepoxide-d5

Synonym: 7-Chloro-N-methyl-5-(phenyl-::FId::FN5)-3H-1,4-benzodiazepin-2-amine 4-oxide

Purchase

Properties

isotopic purity   98 atom % D
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?