EMAIL THIS PAGE TO A FRIEND
C0580 Sigma

Centricoll

pH 9.0±0.5 (20 °C)

Synonym: Silica particles coated with polyvinylpyrrolidone

Purchase

Properties

Related Categories Cell Biology, Centrifugation Media, Colloidal Silica Media, Core Bioreagents, Density Gradient Media,
sterility   sterile-filtered
pH   9.0±0.5 (20 °C)
osmolality   ~25 mOsm/kg H2O
conductivity   <1.0 mS/cm
viscosity   10 cP±5 cP(lit.)
density   1.1 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Colloidal PVP coated silica for cell separation

Legal Information

Centricoll is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?