EMAIL THIS PAGE TO A FRIEND
C0596 Sigma

Clausen medium

Recommended for sterility testing

Purchase

Properties

Related Categories All Liquid Media/Broths, Media, Microbiology More...

Description

Components

Ingredients (g/L)
Casein enzymaic hydrolysate, 15.00
Papaic digest of soyabean meal, 3.00
Yeast extract, 6.00
Dextrose, 6.00
Sodium chloride, 2.50
Dipotassium phosphate, 2.00
Sodium citrate, 1.00
L-Cystine, 0.50
L-Asparagine, 1.25
Sodium dithionite, 0.40
Sodium thioglycollate, 0.50
Lecithin, 0.30
Magnesium sulphate, 0.40
Calcium chloride, 0.004
Cobalt sulphate, 0.001
Cupric sulphate, 0.001
Ferrous sulphate, 0.001
Zinc sulphate, 0.001
Manganese chloride, 0.002
Resazurin, 0.001
Agar, 0.75

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?