| Related Categories | Cell Culture Supplies, Culture Boxes and Accessories, Labware, Magenta Vessels and Accessories, Plant Biotechnology, |
| material | polypropylene |
| description | Autoclavable and reusable |
Polypropylene coupler for joining two Magenta vessel bodies (Prod. No. V8380 or V8505) in any combination.
Magenta is a trademark of Magenta Corp.