• USA Home
  • C0684 - Chorionic gonadotropin human

EMAIL THIS PAGE TO A FRIEND
C0684 Sigma

Chorionic gonadotropin human

from human pregnancy urine, lyophilized powder, vial of 1,000 IU

Synonym: Choriogonin, HCG

Purchase

Properties

biological source   from human pregnancy urine
sterility   sterile-filtered
form   lyophilized powder
potency   ~14,000 I.U. per mg
mol wt   mol wt 37.9 kDa
packaging   vial of 1,000 IU
drug control   Home Office Schedule 4.2
storage temp.   −20°C
Gene Information   human ... CGB5(93659)

Description

General description

Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?