• USA Home
  • C0761 - Cytochrome c from chicken heart

EMAIL THIS PAGE TO A FRIEND
C0761 Sigma

Cytochrome c from chicken heart

≥95% based on Mol. Wt. 12,222 basis

Purchase

Properties

Related Categories Cell Biology, Cytochrome c, Cytochromes, Electron Transport and Cellular Respiration, Metabolic Pathways,
assay   ≥95% based on Mol. Wt. 12,222 basis
storage temp.   −20°C
Gene Information   chicken ... CYCS(420624)

Description

Biochem/physiol Actions

The ready fluctuation of cytochrome c within the cell between ferrous and ferric states, makes it an efficient biological electron-transporter. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal catalyst of respiration, it forms the essential electron-bridge between the respirable substrates and oxygen.

Other Notes

View more information on cytochrome c and electron transport at www.sigma-aldrich.com/enzymeexplorer.

Preparation Note

Prepared without using TCA.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

C4011 Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?