EMAIL THIS PAGE TO A FRIEND
C0783 Sigma

β-Casomorphin human

≥97% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Opioid Peptides, Other Opioid Peptides,
InChI Key   ADBHAJDGVKLXHK-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... CSN1S1(1446), CSN3(1448)

Description

Amino Acid Sequence

Tyr-Pro-Phe-Val-Glu-Pro-Ile

Biochem/physiol Actions

Opioid peptide first isolated from an enzymatic digest of casein that is an agonist at μ opioid receptors. Casomorphin 1-7 stimulates fat intake in rats.

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?