EMAIL THIS PAGE TO A FRIEND
C0845 Sigma

6Ckine from mouse

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Synonym: Exodus-2, SLC

Purchase

Properties

Related Categories 6Ckine/Exodus/SLC, Cell Biology, Cell Signaling and Neuroscience, Chemokines, Cytokines and Growth Factors,
recombinant   expressed in E. coli
assay   ≥98% (HPLC)
  ≥98% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   −20°C
Gene Information   mouse ... Ccl21a(18829), Ccl21b(20298)

Description

Analysis Note

The biological activity is measured by its ability to chemoattract total murine T cell population.

Physical form

Lyophilized with no additives.

Biochem/physiol Actions

The expression of mouse 6Ckine has been detected at high levels in the spleen and lung. Recombinant mouse 6Ckine chemotattracts in vitro thymocytes and activated T-cells.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?