EMAIL THIS PAGE TO A FRIEND
C0862 Sigma

CGP-7930

>98% (HPLC), solid

Synonym: 3-(3′,5′-Di-tert-butyl-4′-hydroxy)phenyl-2,2-dimethylpropanol

  • Empirical Formula (Hill Notation) C19H32O2

  • Molecular Weight 292.46

  •  MDL number MFCD04040028

  •  PubChem Substance ID 24278293

Purchase

Properties

Related Categories Approved Therapeutics/Drug Candidates, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, GABAergics,
InChI Key   XLWJPQQFJNGUPA-UHFFFAOYSA-N
assay   >98% (HPLC)
form   solid
color   white
solubility   DMSO: soluble22 mg/mL
Gene Information   human ... GABBR1(2550), GABBR2(9568)

Description

Biochem/physiol Actions

Positive allosteric modulator of GABAB receptors.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?