• USA Home
  • C0910 - Cibacron Blue 3GA Agarose

EMAIL THIS PAGE TO A FRIEND
C0910 Sigma

Cibacron Blue 3GA Agarose

Type 100, saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
InChI Key   VZPXDCIISFTYOM-UHFFFAOYSA-K
type   Type 100
form   saline suspension
extent of labeling   0.05-0.2 μmol per mL
matrix   4% beaded agarose
capacity   0.5-3 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?