• USA Home
  • C0931 - Monoclonal Anti-Calmodulin antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C0931 Sigma

Monoclonal Anti-Calmodulin antibody produced in mouse

clone 1F11, ascites fluid

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Intracellular Calcium Signaling, Antibodies to Calcium-Associated Proteins,
species reactivity   rat, bovine, chicken, Dictyostelium discoideum
application(s)   western blot: 1:100 using a bovine brain calmodulin preparation
clone   1F11, monoclonal
antibody form   ascites fluid
isotype   IgG1
contains   15 mM sodium azide as preservative
shipped in   dry ice
storage temp.   −20°C
Gene Information   rat ... Calm1(24242)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

preparation of purified calmodulin from Dictyostelium discoideum conjugated to KLH.

Specificity

The antibody recognizes native and SDS-denatured calmodulin.

These antibodies are directed against different epitopes within the calmodulin molecule.

Application

Monoclonal Anti-Calmodulin antibody produced in mouse is suitable for western blotting at a dilution of 1:100 using a bovine brain calmodulin preparation.

Biochem/physiol Actions

Calmodulin (CaM) is a calcium-binding messenger protein found in all eukaryotic cells. CaM plays an important role in stabilization of structural intermediates and precise positioning of FMN domain. It activates FMN domain to have a flexible movement between the reductase and oxygenase domains. It also helps in rapid electron transfer in the homodimer of eNOS.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?