• USA Home
  • C0995 - CHK2, active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
C0995 Sigma

CHK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: CDS1, CHEK2, HuCds1, LFS2, PP1425, RAD53, RP11-436C9.1

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   654-884 nmol/min·mg
mol wt   mol wt ~88 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CHEK2(11200)

Description

Biochem/physiol Actions

CHK2 is rapidly phosphorylated and activated in response to replication blocks and DNA damage; the response to DNA damage occurs in an ataxia telangiectasia mutated (ATM)-dependent manner. Expression of wild-type CHK2 leads to increased p53 stabilization after DNA damage, whereas expression of a dominant-negative CHK2 mutant abrogated both phosphorylation of p53 on Ser-20 and p53 stabilization.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?