• USA Home
  • C1012 - α-Chymotrypsin, Inactivated from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
C1012 Sigma

α-Chymotrypsin, Inactivated from bovine pancreas

Type VI (Inactivated with DFP), lyophilized powder, <0.2 units/mg protein

Purchase

Properties

Related Categories 3.4.x.x Peptidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Chymotrypsin, alpha-,
type   Type VI (Inactivated with DFP)
form   lyophilized powder
mol wt   mol wt 25 kDa
purified by   3× crystallization
storage temp.   −20°C
Gene Information   cow ... CTRB1(618826)

Description

Analysis Note

Protein determined by A1%/280

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase. It has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, and α2-macroglobulin, 10 mM Cu2+ and Hg2+.

Unit Definition

One unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GC3050000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

A2504 6-Aminocaproic acid, ≥99% (titration), powder
A7824 6-Aminocaproic acid, BioUltra, ≥99%
D0879 Diisopropylfluorophosphate
L2023 Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
T9777 Trypsin-chymotrypsin inhibitor from Glycine max (soybean), lyophilized powder
A1153 Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
P7626 Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
B1552 Bromoenol lactone, ≥98% (TLC)
A9024 α1-Antitrypsin from human plasma, salt-free, lyophilized powder
A6150 α1-Antitrypsin from human plasma, salt-free, lyophilized powder
A9285 α1-Antichymotrypsin from human plasma, ≥92.5% (SDS-PAGE), lyophilized powder
T4376 N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder
C7268 Chymostatin, microbial

substrate

Product #

Description

Add to Cart

A3401 Ala-Ala-Phe-7-amido-4-methylcoumarin, protease substrate
S7388 N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
M4507 4-Methylumbelliferyl p-trimethylammoniocinnamate chloride, powder
S8758 N-Succinyl-Ala-Ala-Phe-7-amido-4-methylcoumarin, protease substrate
A9273 Ala-Ala-Val-Ala p-nitroanilide, protease substrate
S1899 N-Succinyl-Gly-Gly-Phe-p-nitroanilide, protease substrate
S9761 N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin, Leucine aminopeptidase substrate
G3769 Glutaryl-Ala-Ala-Phe-4-methoxy-β-naphthylamide, protease substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?