• USA Home
  • C1035 - Cibacron Blue 3GA Agarose

EMAIL THIS PAGE TO A FRIEND
C1035 Sigma

Cibacron Blue 3GA Agarose

Type 300, saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
InChI Key   VZPXDCIISFTYOM-UHFFFAOYSA-K
type   Type 300
form   saline suspension
extent of labeling   0.2-0.5 μmol per mL
matrix   4% beaded agarose
capacity   1,000-4,000 units/mL binding capacity (lactic dehydrogenase)
  1-5 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?