• USA Home
  • C1124 - CDK3/CyclinE1, active, GST tagged human

EMAIL THIS PAGE TO A FRIEND
C1124 Sigma

CDK3/CyclinE1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: CCNE1

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   17-23 nmol/min·mg
mol wt   CDK3 subunit mol wt ~60 kDa
  cyclin E1 mol wt ~73 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CCNE1(898), CDK3(1018)

Description

Biochem/physiol Actions

CDK3/CyclinE1is a member of the cyclin-dependent protein kinase family and promotes entry into S phase, in part by activating members of the E2F family of transcription factors. CDK3 can also associate with cyclin C and phosphorylates the retinoblastoma 1 protein to promote exit from G0. The CDK3 gene has been mapped to chromosomal location of 17q22-qter, telomeric to the BRCA1 gene by somatic cell hybrids analysis. The presence of a single point mutation in the CDK3 gene from several Mus musculus strains commonly used in the laboratory has been reported.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?