EMAIL THIS PAGE TO A FRIEND
C133 Sigma

Charybdotoxin

  • CAS Number 95751-30-7

  • Empirical Formula (Hill Notation) C176H277N57O55S7

  • Molecular Weight 4295.89

  •  MDL number MFCD00146378

Purchase

Properties

storage temp.   −20°C

Description

Amino Acid Sequence

Glp-Phe-Thr-Asn-Val-Ser-Cys-Thr-Thr-Ser-Lys-Glu-Cys-Trp-Ser-Val-Cys-Gln-Arg-Leu-His-Asn-Thr-Ser-Arg-Gly-Lys-Cys-Met-Asn-Lys-Lys-Cys-Arg-Cys-Tyr-Ser [Disulfide Bridges: 7-28, 13-33, 17-35]

Biochem/physiol Actions

High (BK) or intermediate (IK1) conductance calcium-activated and voltage-gated (Kv1.3) channel blocker.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C7802 Charybdotoxin, ≥90% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?