• USA Home
  • C138 - Cysteamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
C138 Sigma

Cysteamine hydrochloride

Synonym: β-Mercapto­ethylamine hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, 2-Amino­ethane­thiol hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Purchase

Properties

InChI Key   OGMADIBCHLQMIP-UHFFFAOYSA-N
mp   67-71 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3
RTECS 
KJ0200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M6500 Cysteamine hydrochloride, ≥98% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?