EMAIL THIS PAGE TO A FRIEND
C1397 Sigma

6-Chloropurine

crystalline

  • CAS Number 87-42-3

  • Empirical Formula (Hill Notation) C5H3ClN4

  • Molecular Weight 154.56

  •  Beilstein Registry Number 5774

  •  EC Number 201-745-6

  •  MDL number MFCD00075825

  •  PubChem Substance ID 329774882

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   ZKBQDFAWXLTYKS-UHFFFAOYSA-N
form   crystalline
color   yellow
mp   >300 °C (dec.)(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UO7520000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?