• USA Home
  • C1431 - CS-C Medium With Serum for Endothelial Cell Lines

EMAIL THIS PAGE TO A FRIEND
C1431 Sigma

CS-C Medium With Serum for Endothelial Cell Lines

With 10% iron supplemented bovine calf serum, L-glutamine, sodium bicarbonate, and HEPES, without phenol red, liquid, sterile-filtered, suitable for cell culture

Purchase

Properties

Related Categories Cell Culture, Endothelial Cells, Human Umbilical Vein Endothelial Cell (HUVEC) Culture Medium, Primary Human Cell Culture, Specialty Media Systems More...
sterility   sterile-filtered
form   liquid
impurities   endotoxin, tested
suitability   suitable for cell culture
storage temp.   2-8°C

Description

Reconstitution

Should be supplemented with Endothelial Cell Growth Factor (E 9640) and Endothelial Cell Attachment Factor (E 9765) for best results.

Application

CS-C Medium has been optimized for the culture of Human Umbilical Vein Endothelial Cells (HUVEC). It contains 10% Iron supplemented calf serum, Lglutamine, sodium bicarbonate, and HEPES. It does not contain phenol red or antibiotics.

CS-C Medium, Product No. C1431 is one of the cell culture media available from Sigma. The selection of a nutrient medium is strongly influenced by 1] type of cell, 2] type of culture [monolayer, suspension, clonal] and 3] ° of chemical definition necessary. It is important to review the literature and more specific references for recommendations concerning medium, supplementation and physiological parameters required for a given cell.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

T4299 Trypsin-EDTA solution, 1 ×, sterile-filtered, BioReagent, suitable for cell culture, 500 BAEE units porcine trypsin and 180 μg EDTA • 4Na per ml in Dulbecco′s PBS without calcium and magnesium

supplement

Product #

Description

Add to Cart

E9640 Endothelial cell growth factor, 100 ×, sterile-filtered, suitable for cell culture
E9765 Endothelial Cell Attachment Factor (ECAF), liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?