• USA Home
  • C1499 - CDK6/CyclinD3, active, His tagged human

EMAIL THIS PAGE TO A FRIEND
C1499 Sigma

CDK6/CyclinD3, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: CCND3, MGC59692, PLSTIRE

Purchase

Properties

Related Categories Active Kinases, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in baculovirus infected Sf9 cells
grade   PRECISIO® Kinase
assay   ≥70% (SDS-PAGE)
form   buffered aqueous glycerol solution
activity   30-42 nmol/min·mg
mol wt   CDK6 subunit mol wt ~40 kDa
  cyclin D3 mol wt ~35 kDa
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CCND3(896), CDK6(1021)

Description

Biochem/physiol Actions

CDK6 is a member of the cyclin-dependent family of protein kinases that are important regulators of cell cycle progression. CDK6 activity is regulated by the D-type cyclins and members of the INK4 family of CDK inhibitors. The CDK6 kinase activity is detected in mid-G1 phase of the cell cycle and is responsible for the phosphorylation and regulation of the activity of tumor suppressor protein Rb. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivities in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?