• USA Home
  • C1540 - Calcium-Binding Protein Vitamin D-induced from bovine mucosa

EMAIL THIS PAGE TO A FRIEND
C1540 Sigma

Calcium-Binding Protein Vitamin D-induced from bovine mucosa

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Casein and Other Milk Proteins, Proteins and Derivatives More...
form   lyophilized powder
composition   Protein, ~60% Lowry
storage temp.   −20°C

Description

Biochem/physiol Actions

The bovine calcium-binding protein shows a 1:2 binding stoichiometry with Ca2+. The protein appears in the cytoplasm of intestinal enterocytes in response to 1,25-dihydroxycholecalciferol.

Packaging

Package size based on protein content

Preparation Note

A modification of the method of Bryant, D.W. and Andrews, P., Biochem. J., 211, 709 (1983).

Physical form

Lyophilized powder containing salts

Application

Calcium-Binding Protein (CaBP) Vitamin D-induced from bovine mucosa is a transport protein important in the translocation of calcium across epithelial membranes. It may be used in the study of Vitamin D-regulated factors involved in active Ca transport present in the renal and intestinal epithelial cells.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?