• USA Home
  • C1544 - Cell Dissociation Solution Non-enzymatic 1x

EMAIL THIS PAGE TO A FRIEND
C1544 Sigma

Cell Dissociation Solution Non-enzymatic 1x

In PBS without Ca2+ and Mg2+., sterile-filtered, BioReagent, suitable for insect cell culture

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Dissociation, Dissociation, General Stem Cell Biology,
sterility   sterile-filtered
product line   BioReagent
suitability   suitable for insect cell culture
storage temp.   2-8°C

Description

Application

Special non-enzymatic formulation for gently dislodging adherent cell types from plastic or glass surfaces.

Price and Availability


Flow Cytometry Instruments

Filter out uncertainty.
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

supplement

Product #

Description

Add to Cart

M8266 Magnesium chloride, anhydrous, ≥98%
M2643 Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
C5670 Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?